3,4'-Bipyridin-2'-amine 95%

Supplier: Ambeed
Warning

A170665-100MG A170665-1G
77151-266EA 39.14 USD
77151-266 77151-268
3,4'-Bipyridin-2'-amine 95%
3,4'-Bipyridin-2'-amine

Precaución: for Research Use Only

Formula: C₁₀H₉N₃
MDL Number: MFCD10703510
CAS Number: 865604-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR