2,4-Difluoro-6-nitrophenol 97%

Supplier: Ambeed
Danger

A170407-10G A170407-1G A170407-250MG A170407-25G A170407-5G
77151-248EA 180.25 USD
77151-248 77151-250 77151-252 77151-254 77151-256
2,4-Difluoro-6-nitrophenol 97%
2,4-Difluoro-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₃
MW: 175.0900 g/mol
MDL Number: MFCD03094186
CAS Number: 364-31-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR