3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 95%

Supplier: Ambeed
Warning

Synonyms: 2-(9H-Carbazol-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (9H-Carbazol-3-yl)boronic acid Pinacol ester

A169750-1G A169750-25G A169750-5G
77151-222EA 8.24 USD
77151-222 77151-224 77151-226
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 95%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole

Precaución: for Research Use Only

Formula: C₁₈H₂₀BNO₂
MW: 293.17 g/mol
MDL Number: MFCD16996081
CAS Number: 855738-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR