6-Bromo-1H-indazole-3-carbonitrile 97%

Supplier: Ambeed
Warning

A169730-100MG A169730-250MG A169730-1G
77151-218EA 5 USD
77151-218 77151-220 77892-135
6-Bromo-1H-indazole-3-carbonitrile 97%
6-Bromo-1H-indazole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrN₃
MDL Number: MFCD06659785
CAS Number: 885278-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR