Astilbin 97%

Supplier: Ambeed
Danger

A169623-100MG A169623-10MG A169623-1MG A169623-25MG A169623-5MG
77151-208EA 77.25 USD
77151-208 77151-210 77151-212 77151-214 77151-216
Astilbin 97%
Astilbin

Precaución: for Research Use Only

Formula: C₂₁H₂₂O₁₁
MW: 450.39 g/mol
MDL Number: MFCD01632717
CAS Number: 29838-67-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR