4-Ethynyl-1-methyl-1H-pyrazole 98%
Supplier: Ambeed
|
Warning
|
A169181-1G
A169181-250MG
A169181-100MG
77151-200EA
110
USD
77151-200
77151-202
77685-370
4-Ethynyl-1-methyl-1H-pyrazole 98%
4-Ethynyl-1-methyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₆H₆N₂ MW: 106.125 g/mol |
MDL Number:
MFCD17281917 CAS Number: 39806-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...