1-Bromo-2,4-dichlorobenzene 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,4-Dichlorobromobenzene
A169134-100G
A169134-10G
A169134-25G
A169134-500G
A169134-5G
77151-192EA
27
USD
77151-192
77151-194
77151-196
77151-198
77902-005
1-Bromo-2,4-dichlorobenzene 97%
1-Bromo-2,4-dichlorobenzene
Precaución: for Research Use Only
Formula:
C₆H₃BrCl₂ MW: 225.9 |
MDL Number:
MFCD00018333 CAS Number: 1193-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...