trans-1,4-Diaminocyclohexane 97%

Supplier: Ambeed
Danger

A169117-10G A169117-1G A169117-25G A169117-5G A169117-100G
77151-184EA 10 USD
77151-184 77151-186 77151-188 77151-190 77611-648
trans-1,4-Diaminocyclohexane 97%

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MDL Number: MFCD00075174
CAS Number: 2615-25-0
UN: 3259
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR