1-Boc-3-fluoroazetidine-3-methanol 95%

Supplier: Ambeed
Warning

A168556-100MG A168556-1G A168556-250MG A168556-5G
77151-172EA 12 USD
77151-172 77151-174 77151-176 77151-178
1-Boc-3-fluoroazetidine-3-methanol 95%
1-Boc-3-fluoroazetidine-3-methanol

Precaución: for Research Use Only

Formula: C₉H₁₆FNO₃
MDL Number: MFCD17392630
CAS Number: 1126650-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR