Potassium 4-hydroxyphenyltrifluoroborate 97%

Supplier: Ambeed
Warning

A168357-1G A168357-250MG A168357-25G A168357-5G
77151-160EA 46 USD
77151-160 77151-162 77151-164 77151-166
Potassium 4-hydroxyphenyltrifluoroborate 97%
Potassium 4-hydroxyphenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₆H₅BF₃O·K
MDL Number: MFCD09992974
CAS Number: 1015082-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR