4-Octylbenzene-1,3-diol 95%

Supplier: Ambeed
Warning

A168150-1G A168150-250MG
77151-148EA 146.26 USD
77151-148 77151-150
4-Octylbenzene-1,3-diol 95%
4-Octylbenzene-1,3-diol

Precaución: for Research Use Only

Formula: C₁₄H₂₂O₂
MW: 222.32 g/mol
MDL Number: MFCD01684758
CAS Number: 6565-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR