Biotin-PEG4-acid 97%

Supplier: Ambeed
Warning

Synonyms: Biotin-PEG4-propionic acid, 15-(Biotinamido)-4,7,10,13-tetraoxapentadecanoic Acid, PEG4-biotin acid, 17-Oxo-21-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10,13-tetraoxa-16-azahenicosanoic acid

A167407-100MG A167407-50MG A167407-250MG
77151-130EA 144.2 USD
77151-130 77151-132 77567-438
Biotin-PEG4-acid 97%
Biotin-PEG4-acid

Precaución: For research use only.

Formula: C₂₁H₃₇N₃O₈S
MW: 491.60 g/mol
MDL Number: MFCD08457831
CAS Number: 721431-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR