4-Chloro-3-iodobenzaldehyde 95%

Supplier: Ambeed
Warning

A167203-100MG A167203-1G A167203-250MG
77151-124EA 18.54 USD
77151-124 77151-126 77151-128
4-Chloro-3-iodobenzaldehyde 95%
4-Chloro-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClIO
MW: 266.464 g/mol
MDL Number: MFCD09836681
CAS Number: 276866-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR