2-(Difluoromethyl)pyrrolidine hydrochloride 95%

Supplier: Ambeed
Warning

A167107-100MG A167107-250MG
77151-122EA 279 USD
77151-122 77685-359
2-(Difluoromethyl)pyrrolidine hydrochloride 95%
2-(Difluoromethyl)pyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClF₂N
MDL Number: MFCD28501339
CAS Number: 1781041-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR