N-Carbobenzoxyglycine methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: N-Cbz-glycine Methyl Ester, N-Benzyloxycarbonylglycine methyl ester, Z-Gly-OMe

A166758-100G A166758-10G A166758-25G A166758-500G A166758-5G
77151-108EA 97.85 USD
77151-108 77151-110 77151-112 77151-114 77151-116
N-Carbobenzoxyglycine methyl ester 97%
N-Carbobenzoxyglycine methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
MDL Number: MFCD00043631
CAS Number: 1212-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR