3'-Sialyllactose sodium salt 90+% (HPAEC-PAD)

Supplier: Ambeed
Warning

Synonyms: 3'-Sialyllactose sodium, Sodium (2S,4S,5R,6R)-5-acetamido-2-(((2R,3S,4S,5R,6S)-3,5-dihydroxy-2-(hydroxymethyl)-6-(((2R,3R,4R,5R)-1,2,4,5-tetrahydroxy-6-oxohexan-3-yl)oxy)tetrahydro-2H-pyran-4-yl)oxy)-4-hydroxy-6-((1R,2R)-1,2,3-trihydroxypropyl)tetrahydro-2H-pyran-2-carboxylate, Neu5Ac alpha(2-3)Gal beta(1-4)Glc sodium salt

A166609-1G A166609-250MG A166609-100MG A166609-50MG
77151-104EA 459 USD
77151-104 77151-106 77611-626 77685-355
3'-Sialyllactose sodium salt 90+% (HPAEC-PAD)
3'-Sialyllactose sodium salt

Precaución: for Research Use Only

Formula: C₂₃H₃₈NNaO₁₉
MDL Number: MFCD00166999
CAS Number: 128596-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR