Neopentyl benzenesulfonate 97%

Supplier: Ambeed
Danger

Synonyms: Neopentyl Benzenesulphonate

A166099-1G A166099-25G A166099-5G
77151-088EA 21 USD
77151-088 77151-090 77151-092
Neopentyl benzenesulfonate 97%
Neopentyl benzenesulfonate

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃S
MDL Number: MFCD06797068
CAS Number: 75620-67-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR