6-Acetyl-8-cyclopentyl-5-methyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one hydrochloride1 ≥98%

Supplier: Ambeed
Warning

Synonyms: Palbociclib monohydrochloride

A165771-100MG A165771-10MG A165771-250MG A165771-1G A165771-1MG A165771-50MG A165771-5MG
77151-076EA 91.67 USD
77151-076 77151-078 77151-080 77611-614 77685-348 77685-349 77685-350
6-Acetyl-8-cyclopentyl-5-methyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one hydrochloride1 ≥98%
6-Acetyl-8-cyclopentyl-5-methyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one hydrochloride1

Precaución: for Research Use Only

Formula: C₂₄H₃₀ClN₇O₂
MW: 483.99 g/mol
MDL Number: MFCD22420809
CAS Number: 827022-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR