CUDC-101 ≥98%

Supplier: Ambeed
Warning

Synonyms: 7-((4-((3-Ethynylphenyl)amino)-7-methoxyquinazolin-6-yl)oxy)-N-hydroxyheptanamide

A165599-10MG A165599-1MG A165599-5MG A165599-100MG
77151-064EA 30.9 USD
77151-064 77151-066 77151-068 77685-347
CUDC-101 ≥98%
CUDC-101

Precaución: for Research Use Only

Formula: C₂₄H₂₆N₄O₄
MW: 434.49 g/mol
MDL Number: MFCD15528940
CAS Number: 1012054-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR