IDO1 Inhibitor I 98%

Supplier: Ambeed
Warning

A165598-10MG A165598-1MG A165598-5MG
77151-058EA 12 USD
77151-058 77151-060 77151-062
IDO1 Inhibitor I 98%
IDO1 Inhibitor I

Precaución: for Research Use Only

Formula: C₉H₇ClFN₅O₂
MDL Number: MFCD18633286
CAS Number: 914471-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR