(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(1-(tert-butoxycarbonyl)-1H-indol-3-yl)butanoic acid 98+%

Supplier: Ambeed
Warning

A165414-100MG A165414-1G A165414-250MG
77151-052EA 90.64 USD
77151-052 77151-054 77151-056
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(1-(tert-butoxycarbonyl)-1H-indol-3-yl)butanoic acid 98+%
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(1-(tert-butoxycarbonyl)-1H-indol-3-yl)butanoic acid

Precaución: for Research Use Only

Formula: C₃₂H₃₂N₂O₆
MW: 540.60 g/mol
MDL Number: MFCD09842083
CAS Number: 357271-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR