N-Butyl-4-nitroaniline 97%

Supplier: Ambeed
Warning

A164362-1G A164362-5G
77151-024EA 26 USD
77151-024 77151-026
N-Butyl-4-nitroaniline 97%
N-Butyl-4-nitroaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
MDL Number: MFCD00024635
CAS Number: 58259-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR