Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate hydrate(2:1:x) 95%

Supplier: Ambeed
Warning

A164286-100G A164286-10G A164286-25G
77151-018EA 45.32 USD
77151-018 77151-020 77151-022
Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate hydrate(2:1:x) 95%
Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate hydrate(2:1:x)

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₅Na₂O₈P
MW: 409.20 g/mol
CAS Number: 146877-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR