1-((4'-Chloro-5,5-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazine dihydrochloride 97%

Supplier: Ambeed
Warning

A163903-100G A163903-1G A163903-25G A163903-5G A163903-250MG
77151-006EA 156 USD
77151-006 77151-008 77151-010 77151-012 77892-108
1-((4'-Chloro-5,5-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazine dihydrochloride 97%
1-((4'-Chloro-5,5-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₂₉Cl₃N₂
MDL Number: MFCD28334264
CAS Number: 1628047-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR