N,N-Bis-Boc-N-allylamine 98%

Supplier: Ambeed
Warning

A163548-1G A163548-250MG A163548-5G
77151-000EA 86.52 USD
77151-000 77151-002 77151-004
N,N-Bis-Boc-N-allylamine 98%
N,N-Bis-Boc-N-allylamine

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.32 g/mol
MDL Number: MFCD04115302
CAS Number: 115269-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR