4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylidene)aniline

A163447-1G A163447-250MG A163447-5G
77150-990EA 33.99 USD
77150-990 77150-992 77150-994
4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester 98%
4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₄BNO₃
MW: 337.22 g/mol
MDL Number: MFCD10567027
CAS Number: 871366-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR