1,3-Dibromo-2,5-difluorobenzene 97%

Supplier: Ambeed
Warning

A163127-100G A163127-25G A163127-5G
77150-982EA 130 USD
77150-982 77150-984 77150-986
1,3-Dibromo-2,5-difluorobenzene 97%
1,3-Dibromo-2,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₂
MDL Number: MFCD09878120
CAS Number: 128259-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR