Diethyl benzylphosphonate 97%

Supplier: Ambeed
Warning

A162736-100G A162736-10G A162736-1G A162736-25G A162736-500G A162736-5G
77150-958EA 96.82 USD
77150-958 77150-960 77150-962 77150-964 77150-966 77150-968
Diethyl benzylphosphonate 97%
Diethyl benzylphosphonate

Precaución: for Research Use Only

Formula: C₁₁H₁₇O₃P
MW: 228.23 g/mol
MDL Number: MFCD00009078
CAS Number: 1080-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR