2,3-Pyrazinedicarboxylic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Pyrazinecarboxylic anhydride

A162440-25G A162440-5G A162440-100G
77150-954EA 18 USD
77150-954 77150-956 77900-723
2,3-Pyrazinedicarboxylic anhydride 98%
2,3-Pyrazinedicarboxylic anhydride

Precaución: for Research Use Only

Formula: C₆H₂N₂O₃
MDL Number: MFCD00179418
CAS Number: 4744-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR