Benzotriazole 98%

Supplier: Ambeed
Danger

Synonyms: 1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole

A162426-100G A162426-500G
77150-950EA 10 USD
77150-950 77150-952
Benzotriazole 98%
Benzotriazole

Precaución: for Research Use Only

Formula: C₆H₅N₃
MDL Number: MFCD00005699
CAS Number: 95-14-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR