H-Gln-OMe·HCl 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A162369-1G A162369-5G A162369-250MG
77150-940EA 54 USD
77150-940 77592-974 77683-346
H-Gln-OMe·HCl 98%, Ambeed.Inc
H-Gln-OMe·HCl

Precaución: For research use only.

Formula: C₆H₁₂N₂O₃·HCl
MDL Number: MFCD00237059
CAS Number: 32668-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR