VRT752271 99%

Supplier: Ambeed
Warning

Synonyms: Ulixertinib, BVD 523, N-[(1S)-1-(3-chlorophenyl)-2-hydroxyethyl]-4-[5-chloro-2-(propan-2-ylamino)pyridin-4-yl]-1H-pyrrole-2-carboxamide, VRT-752271, VRT752271, BVD-523, (S)-4-(5-Chloro-2-(isopropylamino)pyridin-4-yl)-N-(1-(3-chlorophenyl)-2-hydroxyethyl)-1H-pyrrole-2-carboxamide, BVD523

A162263-100MG A162263-10MG A162263-250MG A162263-1G
77150-932EA 154.5 USD
77150-932 77150-934 77150-936 77685-329
VRT752271 99%
VRT752271

Precaución: for Research Use Only

Formula: C₂₁H₂₂Cl₂N₄O₂
MW: 433.33 g/mol
MDL Number: MFCD22628898
CAS Number: 869886-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR