tert-Butyl (3R,5S)-6-hydroxy-3,5-O-isopropylidene-3,5-dihydroxyhexanoate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl (4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxane-4-acetate, 2-[(4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetic acid tert-Butyl ester, tert-Butyl 2-[(4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate, (4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxane-4-acetic acid tert-Butyl ester
A160432-10G
A160432-1G
A160432-25G
A160432-5G
77150-858EA
62.83
USD
77150-858
77150-860
77150-862
77150-864
tert-Butyl (3R,5S)-6-hydroxy-3,5-O-isopropylidene-3,5-dihydroxyhexanoate 95%
tert-Butyl (3R,5S)-6-hydroxy-3,5-O-isopropylidene-3,5-dihydroxyhexanoate
Precaución: for Research Use Only
Formula:
C₁₃H₂₄O₅ MW: 260.3300 g/mol |
MDL Number:
MFCD23098985 CAS Number: 124655-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...