4-(Benzo[d][1,3]dioxol-5-yl)thiazol-2-amine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Benzo[1,3]dioxol-5-yl-thiazol-2-ylamine

A160404-100MG A160404-1G A160404-250MG
77150-850EA 33.99 USD
77150-850 77150-852 77150-854
4-(Benzo[d][1,3]dioxol-5-yl)thiazol-2-amine 95%
4-(Benzo[d][1,3]dioxol-5-yl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂S
MW: 220.25 g/mol
MDL Number: MFCD01366560
CAS Number: 185613-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR