Fmoc-n-(butyl)-glycine 96%

Supplier: Ambeed
Warning

Synonyms: Fmoc-n-Butyl-Gly-OH

A159508-1G A159508-250MG A159508-5G
77150-814EA 77 USD
77150-814 77150-816 77150-818
Fmoc-n-(butyl)-glycine 96%
Fmoc-n-(butyl)-glycine

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄
MW: 353.41 g/mol
MDL Number: MFCD08457838
CAS Number: 234442-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR