Methyl-p-anisate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxybenzoic acid methyl ester, 4-Anisic acid methyl ester, p-Anisic acid methyl ester, Methyl 4-methoxybenzoate

A159505-1000G A159505-100G A159505-10G A159505-25G A159505-500G
77150-804EA 200.85 USD
77150-804 77150-806 77150-808 77150-810 77150-812
Methyl-p-anisate 97%
Methyl-p-anisate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00008437
CAS Number: 121-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR