1,2,3-Trifluoro-5-nitrobenzene 98%

Supplier: Ambeed
Danger

Synonyms: 1,2,3-Trifluoro-5-nitrobenzene, 3,4,5-Trifluoronitrobenzene

A159467-100G A159467-10G A159467-1G A159467-25G A159467-5G
77150-794EA 97 USD
77150-794 77150-796 77150-798 77150-800 77150-802
1,2,3-Trifluoro-5-nitrobenzene 98%
1,2,3-Trifluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₃NO₂
MDL Number: MFCD00456803
CAS Number: 66684-58-0
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR