H-D-Glu(OBzl)-OBzl·HCl 97%

Supplier: Ambeed
Warning

Synonyms: (R)-Dibenzyl 2-aminopentanedioate hydrochloride

A158719-100G A158719-1G A158719-25G A158719-5G
77150-766EA 73 USD
77150-766 77150-768 77150-770 77150-772
H-D-Glu(OBzl)-OBzl·HCl 97%
H-D-Glu(OBzl)-OBzl·HCl

Precaución: for Research Use Only

Formula: C₁₉H₂₁NO₄·HCl
MDL Number: MFCD00190908
CAS Number: 146844-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR