Sodium (6-((5-fluoro-2-((3,4,5-trimethoxyphenyl)amino)pyrimidin-4-yl)amino)-2,2-dimethyl-3-oxo-2,3-dihydro-4H-pyrido[3,2-b][1,4]oxazin-4-yl)methyl phosphate hexahydrate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fostamatinib disodium hexahydrate
A158498-100MG
A158498-10MG
A158498-50MG
A158498-5MG
A158498-250MG
77150-756EA
232
USD
77150-756
77150-758
77150-760
77150-762
77150-764
77900-811
Sodium (6-((5-fluoro-2-((3,4,5-trimethoxyphenyl)amino)pyrimidin-4-yl)amino)-2,2-dimethyl-3-oxo-2,3-dihydro-4H-pyrido[3,2-b][1,4]oxazin-4-yl)methyl phosphate hexahydrate ≥98%
Sodium (6-((5-fluoro-2-((3,4,5-trimethoxyphenyl)amino)pyrimidin-4-yl)amino)-2,2-dimethyl-3-oxo-2,3-dihydro-4H-pyrido[3,2-b][1,4]oxazin-4-yl)methyl phosphate hexahydrate
Precaución: for Research Use Only
Formula:
C₂₃H₃₆FN₆Na₂O₁₅P |
MDL Number:
MFCD19443649 CAS Number: 914295-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...