(R)-N-Methyl-N-(pyrrolidin-3-yl)acetamide hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A158435-1G
A158435-250MG
77150-752EA
344
USD
77150-752
77150-754
(R)-N-Methyl-N-(pyrrolidin-3-yl)acetamide hydrochloride 98%
(R)-N-Methyl-N-(pyrrolidin-3-yl)acetamide hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₅ClN₂O |
MDL Number:
MFCD29037478 CAS Number: 1788036-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...