(E)-1-(2-Hydroxy-4-methoxy-5-(3-methylbut-2-en-1-yl)phenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one 95%

Supplier: Ambeed
Warning

Synonyms: 4'-O-Methylbavachalcone

A158292-10MG A158292-5MG A158292-1MG
77150-748EA 38.11 USD
77150-748 77150-750 77611-518
(E)-1-(2-Hydroxy-4-methoxy-5-(3-methylbut-2-en-1-yl)phenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one 95%
(E)-1-(2-Hydroxy-4-methoxy-5-(3-methylbut-2-en-1-yl)phenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one

Precaución: for Research Use Only

Formula: C₂₁H₂₂O₄
MW: 338.4 g/mol
MDL Number: MFCD02183383
CAS Number: 20784-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR