tert-Butyl 3-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methyl)azetidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A158175-100MG A158175-50MG A158175-1G A158175-250MG
77150-738EA 74.16 USD
77150-738 77150-740 77685-300 77685-301
tert-Butyl 3-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methyl)azetidine-1-carboxylate 95%
tert-Butyl 3-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methyl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₃₀BN₃O₄
MW: 363.26 g/mol
MDL Number: MFCD18383284
CAS Number: 877399-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR