Methyl 2-(S)-[n-carbobenzyloxy]amino-3-aminopropionate, hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Z-Dap-OMe·HCl, Methyl 2-(S)-[N-Carbobenzyloxy]amino-3-aminopropionate, Hydrochloride

A158095-1G A158095-250MG A158095-5G A158095-25G
77150-732EA 53.56 USD
77150-732 77150-734 77150-736 77549-996
Methyl 2-(S)-[n-carbobenzyloxy]amino-3-aminopropionate, hydrochloride 95%
Methyl 2-(S)-[n-carbobenzyloxy]amino-3-aminopropionate, hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₄·ClH
MW: 288.727 g/mol
MDL Number: MFCD03701202
CAS Number: 35761-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR