4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide 95%

Supplier: Ambeed
Warning

A157844-10G A157844-1G A157844-25G A157844-250MG
77150-726EA 224 USD
77150-726 77150-728 77150-730 77892-078
4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide 95%
4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide

Precaución: for Research Use Only

Formula: C₆H₁₃NO₃S
MDL Number: MFCD00106415
CAS Number: 26475-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR