Methyl 7-methoxy-4-oxo-1,4-dihydroquinoline-6-carboxylate 98%

Supplier: Ambeed
Warning

A157363-1G A157363-5G A157363-25G
77150-712EA 18 USD
77150-712 77150-714 77685-297
Methyl 7-methoxy-4-oxo-1,4-dihydroquinoline-6-carboxylate 98%
Methyl 7-methoxy-4-oxo-1,4-dihydroquinoline-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₄
MDL Number: MFCD13192255
CAS Number: 205448-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR