1,1-Diphenyl-2-propyn-1-ol 97%

Supplier: Ambeed
Warning

A157269-10G A157269-1G A157269-25G A157269-5G A157269-100G
77150-696EA 24 USD
77150-696 77150-698 77150-700 77150-702 77901-801
1,1-Diphenyl-2-propyn-1-ol 97%
1,1-Diphenyl-2-propyn-1-ol

Precaución: for Research Use Only

Formula: C₁₅H₁₂O
MDL Number: MFCD00041570
CAS Number: 3923-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR