7-Bromo-2H-chromen-2-one 95%

Supplier: Ambeed
Warning

A157023-100MG A157023-250MG A157023-1G
77150-692EA 21 USD
77150-692 77150-694 77892-074
7-Bromo-2H-chromen-2-one 95%
7-Bromo-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₉H₅BrO₂
MDL Number: MFCD00152014
CAS Number: 19063-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR