Ganetespib (STA-9090) 98%

Supplier: Ambeed
Warning

A156855-1MG A156855-5MG A156855-250MG A156855-2MG A156855-10MG
77150-684EA 14 USD
77150-684 77150-686 77685-294 77685-295 77892-073
Ganetespib (STA-9090) 98%
Ganetespib (STA-9090)

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₄O₃
MDL Number: MFCD22420818
CAS Number: 888216-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR