BLZ-945 ≥98%

Supplier: Ambeed
Warning

Synonyms: BLZ945, Sotuletinib, 4-((2-(((1R,2R)-2-Hydroxycyclohexyl)amino)benzo[d]thiazol-6-yl)oxy)-N-methylpicolinamide

A156169-10MG A156169-1MG A156169-2MG A156169-5MG A156169-100MG A156169-1G A156169-250MG A156169-50MG
77150-670EA 35 USD
77150-670 77150-672 77150-674 77150-676 77685-285 77685-286 77685-287 77685-288
BLZ-945 ≥98%
BLZ-945

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₄O₃S
MDL Number: MFCD28142668
CAS Number: 953769-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR