4-Ethynylbenzeneboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Ethynyl-phenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane, 2-(4-Ethynyl-phenyl)-4,4,5,5-tetramethyl-[1,3,2]-dioxaborolane

A155711-100MG A155711-1G A155711-250MG A155711-5G
77150-642EA 14 USD
77150-642 77150-644 77150-646 77150-648
4-Ethynylbenzeneboronic acid pinacol ester 95%
4-Ethynylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₇BO₂
MDL Number: MFCD16294504
CAS Number: 1034287-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR